Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377837.1 | NP_001364551.1 | 12 - 13 | 2872 | 121 | |
NM_063636.1 | NP_496037.1 | 12 - 13 | 2878 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |