Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063656.5 | NP_496057.1 | 5 - 6 | 1125 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |