Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063658.8 | NP_496059.2 | 4 - 5 | 478 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |