Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063709.3 | NP_496110.1 | 5 - 6 | 1333 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |