Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063737.2 | NP_496138.1 | 1 - 2 | 212 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |