Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lec-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027361.4 | NP_001022532.1 | 6 - 7 | 852 | 122 | |
NM_001027362.7 | NP_001022533.1 | 6 - 7 | 822 | 122 | ||
NM_001276739.3 | NP_001263668.1 | 7 - 8 | 646 | 122 | ||
NM_063758.8 | NP_496159.1 | 7 - 8 | 681 | 122 | ||
GenBank mRNA | AB038504.1 | - | 6 - 7 | 588 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |