Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026968.4 | NP_001022139.1 | 9 - 10 | 1365 | 97 | |
NM_001026969.4 | NP_001022140.1 | 10 - 11 | 1439 | 97 | ||
NM_001267307.4 | NP_001254236.1 | 10 - 11 | 1452 | 97 | ||
NM_001267308.3 | NP_001254237.1 | 6 - 7 | 1032 | 97 | ||
NM_001306556.3 | NP_001293485.1 | 6 - 7 | 914 | 97 | ||
GenBank mRNA | AF226156.1 | - | 10 - 11 | 1462 | 97 | |
AF233652.1 | - | 9 - 10 | 1387 | 97 | ||
EU812221.1 | - | 6 - 7 | 1032 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |