Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026969.4 | NP_001022140.1 | 4 - 5 | 517 | 66 | |
NM_001267307.4 | NP_001254236.1 | 4 - 5 | 530 | 66 | ||
GenBank mRNA | AF226156.1 | - | 4 - 5 | 540 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |