Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026968.4 | NP_001022139.1 | 5 - 6 | 601 | 76 | |
NM_001026969.4 | NP_001022140.1 | 6 - 7 | 675 | 76 | ||
NM_001267307.4 | NP_001254236.1 | 6 - 7 | 688 | 76 | ||
NM_001267308.3 | NP_001254237.1 | 2 - 3 | 268 | 76 | ||
NM_001306556.3 | NP_001293485.1 | 2 - 3 | 150 | 76 | ||
GenBank mRNA | AF226156.1 | - | 6 - 7 | 698 | 76 | |
AF233652.1 | - | 5 - 6 | 623 | 76 | ||
EU812221.1 | - | 2 - 3 | 268 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |