Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063794.1 | NP_496195.1 | 2 - 3 | 380 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |