Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063834.5 | NP_496235.2 | 4 - 5 | 758 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |