Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cut-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063841.3 | NP_496242.2 | 3 - 4 | 500 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |