Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063855.1 | NP_496256.1 | 1 - 2 | 47 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |