Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063855.1 | NP_496256.1 | 8 - 9 | 1092 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |