Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lact-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063898.4 | NP_496299.1 | 2 - 3 | 451 | 106 |
Gene Symbol: M28.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381581.2 | NP_001366718.1 | 4 - 5 | 1251 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |