Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063923.6 | NP_496324.1 | 2 - 3 | 154 | 59 | |
NM_063924.6 | NP_496325.1 | 2 - 3 | 154 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |