Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mix-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063930.8 | NP_496331.1 | 10 - 11 | 2647 | 90 | |
GenBank mRNA | U96387.1 | - | 10 - 11 | 2509 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |