Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063957.3 | NP_496358.1 | 3 - 4 | 393 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |