Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063990.5 | NP_496391.1 | 5 - 6 | 789 | 85 | |
GenBank mRNA | AF008590.1 | - | 5 - 6 | 749 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |