Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361877.3 | NP_001348695.1 | 9 | 1204 | 67 | |
NM_064022.5 | NP_496423.1 | 8 - 9 | 1152 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |