Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026729.4 | NP_001021900.1 | 13 - 14 | 2019 | 103 | |
NM_001377875.3 | NP_001364585.1 | 13 - 14 | 2097 | 103 | ||
NM_001383953.1 | 12 - 13 | 1907 | 103 | |||
GenBank mRNA | AF173171.1 | - | 12 - 13 | 1929 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |