Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064059.2 | NP_496460.2 | 4 - 5 | 748 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |