Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377878.3 | NP_001364646.1 | 11 - 12 | 6963 | 77 | |
NM_001377879.1 | NP_001364592.1 | 11 - 12 | 7034 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |