Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377877.2 | NP_001364647.1 | 15 - 16 | 8368 | 66 | |
NM_001377878.3 | NP_001364646.1 | 15 - 16 | 8401 | 66 | ||
NM_001377879.1 | NP_001364592.1 | 15 - 16 | 8472 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |