Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377877.2 | NP_001364647.1 | 19 - 20 | 10150 | 113 | |
NM_001377878.3 | NP_001364646.1 | 19 - 20 | 10183 | 113 | ||
NM_001377879.1 | NP_001364592.1 | 19 - 20 | 10254 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |