Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267381.2 | NP_001254310.1 | 3 - 4 | 626 | 65 | |
GenBank mRNA | AB023424.1 | - | 3 - 4 | 618 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |