Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267381.2 | NP_001254310.1 | 6 - 7 | 1638 | 108 | |
NM_001267382.3 | NP_001254311.1 | 1 - 2 | 96 | 108 | ||
GenBank mRNA | AB023424.1 | - | 6 - 7 | 1630 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |