Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dna-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064115.4 | NP_496516.1 | 8 - 9 | 2310 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |