Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267396.3 | NP_001254325.1 | 3 - 4 | 642 | 71 | |
GenBank mRNA | U81043.1 | - | 3 - 4 | 648 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |