Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027242.3 | NP_001022413.1 | 5 - 6 | 1967 | 59 | |
NM_001348640.3 | NP_001335541.1 | 2 - 3 | 707 | 59 | ||
NM_001381624.1 | NP_001368630.1 | 5 - 6 | 1463 | 59 | ||
NM_001383965.1 | 10 - 11 | 2223 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |