Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267406.4 | NP_001254335.1 | 1 - 2 | 41 | 74 | |
NM_001267407.4 | NP_001254336.1 | 1 - 2 | 199 | 74 | ||
NM_001381625.2 | NP_001367160.1 | 2 - 3 | 282 | 74 | ||
NM_001381626.2 | NP_001368162.1 | 1 - 2 | 196 | 74 | ||
NM_064193.4 | NP_496594.2 | 2 - 3 | 307 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |