Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064214.2 | NP_496615.2 | 2 - 3 | 744 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |