Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064227.2 | NP_496628.1 | 2 - 3 | 775 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |