Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027267.5 | NP_001022438.1 | 5 - 6 | 835 | 80 | |
NM_001373808.1 | NP_001359643.1 | 5 - 6 | 830 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |