Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064340.8 | NP_496741.1 | 7 - 8 | 1079 | 99 | |
NM_064341.7 | NP_496742.1 | 5 - 6 | 904 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |