Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267435.3 | NP_001254364.1 | 2 - 3 | 330 | 59 | |
NM_064375.8 | NP_496776.1 | 3 - 4 | 582 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |