Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064459.2 | NP_496860.1 | 1 - 2 | 200 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |