Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmjd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064568.5 | NP_496969.2 | 3 - 4 | 1543 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |