Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377900.1 | NP_001364609.1 | 14 - 15 | 5169 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |