Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mog-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064626.5 | NP_497027.1 | 5 - 6 | 2469 | 69 | |
GenBank mRNA | AF286900.1 | - | 5 - 6 | 2485 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |