Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027317.8 | NP_001022488.1 | 2 - 3 | 484 | 95 | |
NM_001267510.3 | NP_001254439.1 | 2 - 3 | 483 | 95 | ||
NM_001267511.3 | NP_001254440.1 | 2 - 3 | 393 | 95 | ||
NM_001267513.3 | NP_001254442.1 | 2 - 3 | 484 | 95 | ||
NM_001267514.4 | NP_001254443.1 | 2 - 3 | 393 | 95 | ||
NM_001267515.3 | NP_001254444.1 | 2 - 3 | 393 | 95 | ||
NM_001267516.3 | NP_001254445.1 | 2 - 3 | 393 | 95 | ||
NM_001267517.3 | NP_001254446.1 | 2 - 3 | 393 | 95 | ||
NM_001350388.3 | NP_001337294.1 | 12 | 2760 | 95 | ||
NM_001377905.1 | NP_001364664.1 | 12 - 13 | 2920 | 95 | ||
NM_064643.10 | NP_497044.3 | 2 - 3 | 393 | 95 | ||
NM_064644.6 | NP_497045.1 | 2 - 3 | 483 | 95 | ||
NM_064645.6 | NP_497046.1 | 2 - 3 | 484 | 95 | ||
NM_064646.7 | NP_497047.1 | 2 - 3 | 484 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |