Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027640.5 | NP_001022811.1 | 7 - 8 | 1195 | 72 | |
NM_001381690.1 | NP_001367960.1 | 10 - 11 | 1496 | 72 | ||
NM_001384006.1 | 6 - 7 | 1612 | 72 | |||
NM_064789.5 | NP_497190.1 | 12 - 13 | 1998 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |