Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: far-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064872.7 | NP_497273.2 | 3 - 4 | 605 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |