Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064903.3 | NP_497304.1 | 3 - 4 | 825 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |