Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064910.8 | NP_497311.2 | 22 - 23 | 3519 | 54 | |
GenBank mRNA | AJ276028.1 | - | 21 - 22 | 3349 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |