Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027532.5 | NP_001022703.1 | 19 - 20 | 2570 | 134 | |
NM_064910.8 | NP_497311.2 | 23 - 24 | 3641 | 134 | ||
GenBank mRNA | AJ276028.1 | - | 22 - 23 | 3471 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |