Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uaf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027796.4 | NP_001022967.1 | 5 - 6 | 1042 | 69 | |
GenBank mRNA | U79157.1 | - | 5 - 6 | 1042 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |