Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027459.6 | NP_001022630.2 | 4 - 5 | 406 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |