Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sms-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330977.3 | NP_001317879.1 | 5 | 988 | 135 | |
NM_001330978.3 | NP_001317880.1 | 4 - 5 | 793 | 135 | ||
NM_001330979.3 | NP_001317881.1 | 4 - 5 | 835 | 135 | ||
NM_001330980.3 | NP_001317882.1 | 1 - 2 | 217 | 135 | ||
NM_065024.5 | NP_497425.1 | 6 - 7 | 1219 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |