Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pvf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065060.4 | NP_497461.1 | 5 - 6 | 857 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |